Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Boron trifluoride diethyl ether complex (≥46,5% BF₃)

Toxic
Health
Corrosive
Flammable
Boron trifluoride diethyl ether complex (≥46,5% BF₃)
Certificates
Boron trifluoride diethyl ether complex (≥46,5% BF₃)
Boron trifluoride diethyl ether complex (≥46,5% BF₃)

2 Options Available Below

Product Details & Documents

CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Physical Form: Liquid Density: 1.130 Refractive Index: 1.3440 Sensitivity: Moisture Sensitive Odour: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194 Packaged under Argon in resealable ChemSeal™ bottles

Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:14,1350
  • Boiling Pt:125…126 °C
  • Melting Pt:–60 °C
  • Density:1.130
  • Flash Pt:47 °C (116 °F)
  • Storage Temperature:Ambient
  • UN:2604
  • ADR:8,I
  •  :Packaged under argon in resealable ChemSeal™ bottles.
  • EINECS:203-689-8
Product Family Options

Product Information