Order Entry
Germany
Orders LinkContactUsLinkComponent
Boron trifluoride diethyl ether complex (∼48% BF₃)

Boron trifluoride diethyl ether complex (∼48% BF₃)

Flammable
Corrosive
Irritant
Health
Boron trifluoride diethyl ether complex (∼48% BF₃)
Certificates
Boron trifluoride diethyl ether complex (∼48% BF₃)
Boron trifluoride diethyl ether complex (∼48% BF₃)

4 Options Available Below

Product Details & Documents
Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:13,01340
  • Boiling Pt:126 °C (760 mmHg)
  • Melting Pt:–60 °C
  • Density:1.12 g/cm³
  • Flash Pt:58 °C
  • Storage Temperature:Ambient
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to straw yellow Liquid (May darken on storage)
  • Refractive index:
    1.3429 to 1.3449 (20°C, 589 nm)
  • Assay:
    47.5 to 49 % (BF3)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,60 €
    21,60 €
    1 * 25 g
       21,60 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 34,80 €
    34,80 €
    1 * 100 g
       34,80 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 130,00 €
    130,00 €
    1 * 1 kg
       130,00 €

  • Glass bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 253,00 €
    253,00 €
    1 * 2,5 kg
       253,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.