Specifications
- Pk:1 KIT
- Description:Amyloid plaque staining reagent
Specifications
About this item
Amylo-Glo RTD reagent is designed to stain amyloid plaques in tissue sections.
- For Amyloid plaques - Both intraneuronal and vascular
- Very bright blue UV excitable stain under physiological conditions
- Unique excitation/emission profile and mild staining
- Combination for multiple immunofluorescent labeling studies
- Storage Up to 6 months after date of receipt at 4 °C protected from light
Amylo-Glo RTD is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and particularly good for confocal and multiple labeling because of its high fluorescent intensity and resistance to photo-bleaching. Moreover because Amylo-Glo® fluoresces in the UV channel, double and triple labeling experiments can be performed very easily.
Amylo-Glo® RTD™ is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and it is particularly good for confocal and multiple labeling because of its high fluorescent intensity and high resistance to photo-bleaching. Moreover because Amylo-Glo® fluoresces in the UV channel, double and triple labeling experiments can be performed very easily - See protocol.