Naphthalene-[D₈] (99% D)
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
CAS Number:1146-65-2
:
Formula:C₁₀D₈
MDL Number:MFCD00001743
:136.11 g/mol
Melting Pt:81…83 °C
Flash Pt:78 °C
EINECS:214-552-7
UN:1334
ADR:4.1,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient