Order Entry
Germany
ContactUsLinkComponent
Deuterium oxide
Deuterium oxide
Catalog # LBCLD2O10
Supplier:  LABC LABORTECHNIK
CAS Number:  
Deuterium oxide
Catalog # LBCLD2O10
Supplier:  LABC LABORTECHNIK
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :20 g/mol
Merck Index:12,02984
Boiling Pt:101.3 °C (1013 hPa)
Melting Pt:3.8 °C
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:232-148-9
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636