Deuterium oxide 100.0 atom % D for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-20-0
:
Formula:D₂O
MDL Number:MFCD00044636
:20 g/mol
Merck Index:12,02984
Boiling Pt:101.3 °C (1013 hPa)
Melting Pt:3.8 °C
Density:1.105 g/cm³
Storage Temperature:Refrigerator
EINECS:232-148-9
Specification Test Results
- Appearance:Clear colorless Liquid
- D-Enrichment:≥99.95 %
- Melting point:150 to 156 °C
- HPLC:≥96.0 %