Order Entry
Germany
ContactUsLinkComponent
Deuterium oxide 99.8 atom % D for NMR spectroscopy
Deuterium oxide 99.8 atom % D for NMR spectroscopy
Certificates
undefined
Deuterium oxide 99.8 atom % D for NMR spectroscopy
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • D-Enrichment:
    ≥99.75 %
  • Melting point:
    69 to 77 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Certificates
Search by Lot Number