Nitrobenzene-[D₅]
Catalog # EHERC15557100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:128.07 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:5 °C
Density:1.25 g/cm³ (20 °C)
Flash Pt:88 °C
Storage Temperature:Ambient
EINECS:224-014-3
UN:1662
ADR:6.1,II
CAS Number:4165-60-0
Formula:C₆D₅NO₂
MDL Number:MFCD00044415