Order Entry
Germany
ContactUsLinkComponent
Pyridine-[D₅] 100.0 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Irritant
Flammable
Pyridine-[D₅] 100.0 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Certificates
undefined
Irritant
Flammable
Pyridine-[D₅] 100.0 atom % D + 0.03% (v/v) TMS for NMR spectroscopy

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.95 %
  • TMS:
    0.03 %

Specification Test Results

Related Information