Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl sulphoxide-[D6]
Dimethyl sulphoxide-[D₆], Sigma-Aldrich®
Dimethyl sulphoxide-[D₆], Sigma-Aldrich®
:
Merck
Dimethyl sulphoxide-[D₆], Sigma-Aldrich®
:
Merck
:
2206-27-1
:
:
D₃CS(O)CD₃
:
MFCD00002090
:
84.09 g/mol
:
189 °C (5 mmHg)
:
20.2 °C
:
1.19 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
82 °C (closed cup)
:
Ambient
:
218-617-0