Order Entry
Germany
ContactUsLinkComponent
 
Petroleum spirit, 80…110 °C
Catalog # LBCL4623500
Supplier:  LABC LABORTECHNIK
CAS Number:  
Petroleum spirit, 80…110 °C
Catalog # LBCL4623500
Supplier:  LABC LABORTECHNIK
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Boiling Pt:80…110 °C (1013 hPa)
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849