Petroleum spirit, 40…60 °C, Suprasolv® hypergrade for organic trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:40…60 °C (1013 hPa)
Density:0.65 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Boiling range (40-60°C):≥ 90 % (v/v)
- Evaporation residue:≤ 3.0 mg/l
- Water:≤ 0.005 %
- Color:≤ 10 Hazen
- GC/ECD (retention range 1,2,4-tri- chlorobenzene to decachlorobiphenyle, individual signals (lindane standard)):≤ 2 pg/ml
- GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)):≤ 2 ng/ml
- GC/MSD (retention range n-undecane to n-tetracontane, scaning area 30 - 600 amu, individual signals (n- tetradecane standard)):≤ 2 ng/ml
Specification Test Results
Related Information
Merck SupraSolv® High Purity Solvents for Gas Chromatography
Product Literature
ProductPage_116740