Molekularsieb 3A (0,3 nm, 3 Å), Perlen 4-8 mesh, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00147627
Melting Pt:400 °C
Density:2 g/cm³
Storage Temperature:Raumtemperatur
Specification Test Results
- Particle Size:4-8 mesh
- Grain size 4 - 8 mesh:Min. 95 %
- Water adsorption capacity:Min. 19.5 %
Specification Test Results
Frequently Bought Together