Order Entry
Germany
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å), rods, Ø ~1,6 mm, Supelco®
Molecular sieve 3A (0.3 nm, 3 Å), rods, Ø ~1,6 mm, Supelco®
Supplier:  Merck
Certificates
undefined
Molecular sieve 3A (0.3 nm, 3 Å), rods, Ø ~1,6 mm, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Identity:
    Conforms
  • Loss on ignition (950 °C):
    ≤10 %
  • Water absorption capacity (24 hrs.,80 % relative humidity):
    ≥15%

Specification Test Results

Frequently Bought Together