Order Entry
Germany
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å)
Molecular sieve 3A (0.3 nm, 3 Å)
Catalog # BIOT005546-1000
Supplier:  BIONET RESEARCH
CAS Number:  
Molecular sieve 3A (0.3 nm, 3 Å)
Catalog # BIOT005546-1000
Supplier:  BIONET RESEARCH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
CAS Number:308080-99-1
 :1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613