Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Soda lime
Soda lime indicator
Soda lime indicator
Corrosive
Soda lime indicator
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Corrosive
Soda lime indicator
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
8006-28-8
:
MDL Number:
MFCD00134094
Merck Index:
14,8569
Storage Temperature:
Ambient
UN:
1907
ADR:
8,III
:
GHS05
Related Information
Documentation
SDS
msds_ALFAA19352
Videos
Thermo Scientific Acroseal 25 Years
Documentation