Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1,4-Cyclohexanediol
1,4-Cyclohexanediol cis- and trans-mixture ≥98%
1,4-Cyclohexanediol cis- and trans-mixture ≥98%
:
Thermo Fisher Scientific
1,4-Cyclohexanediol cis- and trans-mixture ≥98%
:
Thermo Fisher Scientific
:
556-48-9
:
:
C₆H₁₂O₂
:
MFCD00001448
:
116.16 g/mol
:
149…151 °C (20 mmHg)
:
98…103 °C
:
1.17 g/cm³ (21.85 °C)
:
65 °C (149 °F)
:
Ambient
:
209-126-2
msds_ALFAB20446
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years