Order Entry
Germany
ContactUsLinkComponent
Soda lime, with indicator, Sigma-Aldrich®
Corrosive
Soda lime, with indicator, Sigma-Aldrich®
Catalog # SIAL72073-1KG
Supplier:  Merck
CAS Number:  
Corrosive
Soda lime, with indicator, Sigma-Aldrich®
Catalog # SIAL72073-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Storage Temperature:Ambient
UN:1907
ADR:8,III
 :GHS05
CAS Number:8006-28-8
MDL Number:MFCD00134094