

Soda lime, with indicator
Supplier: Merck
Certificates
About this item
CAS Number:8006-28-8
:
MDL Number:MFCD00134094
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:1907
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMERC1.06733.0501_TDS_EN