Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Platinum
Platinum, Sigma-Aldrich®
Platinum, Sigma-Aldrich®
:
Merck
Platinum, Sigma-Aldrich®
:
Merck
:
7440-06-4
:
:
Pt
:
MFCD00011179
:
195.08 g/mol
:
12,07689
:
3827 °C (1013 hPa)
:
1769 °C
:
21.447 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
Ambient
:
231-116-1