Order Entry
Germany
Orders LinkContactUsLinkComponent
Platinum, Sigma-Aldrich®

Platinum, Sigma-Aldrich®

Glass bottle for solids, 1,5 g
Catalog # SIAL303798-1.5G
Supplier:   Merck
Platinum, Sigma-Aldrich®
Catalog # SIAL303798-1.5G
Supplier:  Merck
CAS Number:  7440-06-4
Platinum, Sigma-Aldrich®

Glass bottle for solids, 1,5 g

CAS Number:  7440-06-4
Price
596,00 €
Log in for availability
1 * 1,5 g
Product Details & Documents
Synonyms:  Platinium
  • CAS Number:7440-06-4
  • Formula:Pt
  • MDL Number:MFCD00011179
  • Molecular Weight:195.08 g/mol
  • Merck Index:12,07689
  • Boiling Pt:3827 °C (1013 hPa)
  • Melting Pt:1769 °C
  • Density:21.447 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:231-116-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1,5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.