Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Platinum
Platinum ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Platinum ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
Platinum ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
:
7440-06-4
:
:
Pt
:
MFCD00011179
:
195.08 g/mol
:
12,07689
:
3827 °C (1013 hPa)
:
1769 °C
:
21.447 g/cm³ (20 °C)
:
Ambient
:
231-116-1
msds_ALFA043276
msds_ALFA043276