Order Entry
Germany
ContactUsLinkComponent
Platinum ≥99.95% (metals basis), thin foil, hard, Thickness: 0.00125 mm (0.00005 in)
Platinum ≥99.95% (metals basis), thin foil, hard, Thickness: 0.00125 mm (0.00005 in)
Catalog # 41632.FF
CAS Number:  
Platinum ≥99.95% (metals basis), thin foil, hard, Thickness: 0.00125 mm (0.00005 in)
Catalog # 41632.FF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1643
  • Pk Info:
    25×25 mm

Specifications

About this item

CAS Number:7440-06-4
 :
Formula:Pt
MDL Number:MFCD00011179
 :195.08 g/mol
Merck Index:12,07689
Boiling Pt:3827 °C (1013 hPa)
Melting Pt:1769 °C
Density:21.447 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1