ContactUsLinkComponent
Platinum ≥99.95% (metals basis), wire, hard, Ø 0.368 mm (0.0145 in)
Platinum ≥99.95% (metals basis), wire, hard, Ø 0.368 mm (0.0145 in)
Certificates
Platinum ≥99.95% (metals basis), wire, hard, Ø 0.368 mm (0.0145 in)