Order Entry
Germany
ContactUsLinkComponent
Silica gel nanoparticles 40% (w/w), colloidal dispersion in water, LUDOX® AS-40, Sigma-Aldrich®
Silica gel nanoparticles 40% (w/w), colloidal dispersion in water, LUDOX® AS-40, Sigma-Aldrich®
Supplier:  Merck
Certificates
Silica gel nanoparticles 40% (w/w), colloidal dispersion in water, LUDOX® AS-40, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Identity:
    Confirmed
  • pH at 25 °C:
    9.0 - 9.5
  • Specific gravity (at 60 °F):
    1.286 - 1.300
  • Viscosity (at 25 °C):
    ≤20 cps
  • Silica:
    39.5 - 41.0%
  • Ratio SiO₂/NiO₂:
    160 - 270
  • Surface area:
    129 - 155 m²/g
  • Sulphate (SO₄) as (Na₂SO₄):
    ≤0.02%
  • Assay (T):
    ≥20%
  • Sodium (Na):
    ≤0.10%
  • Particle size:
    20.0 - 24.0 nm
  • Chloride content:
    ≤0.01%

Specification Test Results

Frequently Bought Together