Nickel, powder, Sigma-Aldrich®
Catalog # SIAL268283-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:58.69 g/mol
Boiling Pt:2371 °C (1013 hPa)
Melting Pt:1455 °C
Density:8.9 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-111-4
UN:3089
ADR:4.1,II
:GHS08
CAS Number:7440-02-0
Formula:Ni
MDL Number:MFCD00011137