Nickel, powder, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
:58.69 g/mol
ADR:4.1,II
:M-Clarity™ quality level = MQ100
EINECS:231-111-4
UN:3089
:GHS08
Storage Temperature:Ambient