Order Entry
Germany
ContactUsLinkComponent
Nickel ≥99.99% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Nickel ≥99.99% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40875.KS
CAS Number:  
Nickel ≥99.99% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40875.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-02-0
 :
Formula:Ni
MDL Number:MFCD00011137
 :58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-111-4
UN:3089
ADR:4.1,III
 :GHS08
 :GHS07