


Nickel ≥99.999% (metals basis), powder, Puratronic® (max. 100ppm C)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
:58.69 g/mol
Boiling Pt:2371 °C (1013 hPa)
Melting Pt:1455 °C
Density:8.9 g/cm³ (25 °C)
EINECS:231-111-4
UN:3089
ADR:4.1,II
:GHS08
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together