Order Entry
Germany
ContactUsLinkComponent
Nickel ≥99.995% (metals basis), rods, Puratronic®, Ø 12 mm (0.47 in)
Nickel ≥99.995% (metals basis), rods, Puratronic®, Ø 12 mm (0.47 in)
Catalog # 11499.HF
CAS Number:  
Nickel ≥99.995% (metals basis), rods, Puratronic®, Ø 12 mm (0.47 in)
Catalog # 11499.HF
CAS Number:  

Specifications

  • Pk:
    10 mm
  • Seq:
    1622

Specifications

About this item

 :
 :58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-111-4
UN:3089
ADR:4.1,III
 :GHS08
 :GHS07
CAS Number:7440-02-0
Formula:Ni
MDL Number:MFCD00011137