Order Entry
Germany
ContactUsLinkComponent
Nickel 99.97% (trace metals basis), spheres 5 --> 11 mm
Nickel 99.97% (trace metals basis), spheres 5 --> 11 mm
Catalog # ACRO201672500
CAS Number:  
undefined
Nickel 99.97% (trace metals basis), spheres 5 --> 11 mm
Catalog # ACRO201672500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    Spheres
  • Assay:
    ≥99.92 % (Ni)

Specification Test Results

About this item

 :
 :58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1455 °C
Density:8,9 g/cm³ (20 °C)
EINECS:231-111-4
ADR:4.1,III
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:7440-02-0
Formula:Ni
MDL Number:MFCD00011137