Order Entry
Germany
ContactUsLinkComponent
Tungsten, powder, Sigma-Aldrich®
Tungsten, powder, Sigma-Aldrich®
Catalog # SIAL357421-10G
Supplier:  Merck
CAS Number:  
Tungsten, powder, Sigma-Aldrich®
Catalog # SIAL357421-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :183.84 g/mol
ADR:4.1,II
 :M-Clarity™ quality level = MQ100
EINECS:231-143-9
UN:3089
CAS Number:7440-33-7
Formula:W
MDL Number:MFCD00011461