Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tungsten
Tungsten, powder, Sigma-Aldrich®
Tungsten, powder, Sigma-Aldrich®
Tungsten, powder, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Tungsten, powder, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7440-33-7
:
Formula:
W
MDL Number:
MFCD00011461
:
183.84 g/mol
ADR:
4.1,II
:
M-Clarity™ quality level = MQ100
EINECS:
231-143-9
UN:
3089