Order Entry
Germany
ContactUsLinkComponent
Tungsten, powder, Sigma-Aldrich®
Tungsten, powder, Sigma-Aldrich®
Catalog # 1.12406.0100
Supplier:  Merck
CAS Number:  
Tungsten, powder, Sigma-Aldrich®
Catalog # 1.12406.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    3370 °C
  • Molar Mass:
    183.85 g/mol
  • Bulk Density:
    4000 kg/m3
  • Boiling Point:
    5930 °C

Specification Test Results

About this item

 :
 :183.84 g/mol
ADR:4.1,II
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-143-9
UN:3089
CAS Number:7440-33-7
Formula:W
MDL Number:MFCD00011461