Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tungsten
Tungsten ≥99.9% (metals basis), powder APS 12 µm
Tungsten ≥99.9% (metals basis), powder APS 12 µm
:
Thermo Fisher Scientific
Tungsten ≥99.9% (metals basis), powder APS 12 µm
:
Thermo Fisher Scientific
:
7440-33-7
:
:
W
:
MFCD00011461
:
183.84 g/mol
:
Ambient
:
231-143-9
:
3089
:
4.1,III
:
GHS02
msds_ALFA010401
Thermo Scientific Acroseal 25 Years