Tungsten, Sigma-Aldrich®
Catalog # SIAL267554-9.5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:9,5 g
- Seq:3233
Specifications
About this item
:
:183.84 g/mol
Merck Index:12,09945
Boiling Pt:5660 °C (1013 hPa)
Melting Pt:3410 °C
Density:19 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:231-143-9
CAS Number:7440-33-7
Formula:W
MDL Number:MFCD00011461