Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tungsten
Tungsten, gauze woven from wire 100 mesh, Ø 0.0254 mm (0.001 in)
Tungsten, gauze woven from wire 100 mesh, Ø 0.0254 mm (0.001 in)
:
Thermo Fisher Scientific
Tungsten, gauze woven from wire 100 mesh, Ø 0.0254 mm (0.001 in)
:
Thermo Fisher Scientific
:
7440-33-7
:
:
W
:
MFCD00011461
:
183.84 g/mol
:
12,09945
:
5660 °C (1013 hPa)
:
3410 °C
:
19 g/cm³ (20 °C)
:
Ambient
:
231-143-9
msds_ALFA040924
Thermo Scientific Acroseal 25 Years