Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tungsten
Tungsten ≥99.95% (metals basis), wire, Ø 0.1 mm (0.004 in)
Tungsten ≥99.95% (metals basis), wire, Ø 0.1 mm (0.004 in)
:
Thermo Fisher Scientific
Tungsten ≥99.95% (metals basis), wire, Ø 0.1 mm (0.004 in)
:
Thermo Fisher Scientific
:
7440-33-7
:
:
W
:
MFCD00011461
:
183.84 g/mol
:
12,09945
:
5660 °C (1013 hPa)
:
3410 °C
:
19 g/cm³ (20 °C)
:
Ambient
:
231-143-9
msds_ALFA010404