0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Titanium
Titanium ≥99% (metals basis), powder -325 mesh
Titanium ≥99% (metals basis), powder -325 mesh
:
Thermo Fisher Scientific
Titanium ≥99% (metals basis), powder -325 mesh
:
Thermo Fisher Scientific
:
7440-32-6
:
:
Ti
:
MFCD00011264
:
47.88 g/mol
:
3277 °C (1013 hPa)
:
1677 °C
:
4.5 g/cm³ (20 °C)
:
Ambient
:
231-142-3
:
2546
:
4.2,II
:
GHS02
msds_ALFA010386
Thermo Scientific Acroseal 25 Years