ContactUsLinkComponent
Titanium ≥99.98% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Titanium ≥99.98% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
  42388.QK
 :  
Titanium ≥99.98% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
  42388.QK
 :  
  • Pk:
    25
  • Seq:
    0025
 :
 :47.88 g/mol
 :12,09610
 :3262 °C (1013 hPa)
 :1660 °C
 :4.51 g/cm³ (20 °C)
 :Ambient
 :231-142-3
 :GHS02
 :7440-32-6
 :Ti
 :MFCD00011264