Silver, Sigma-Aldrich®
Catalog # SIAL348783-200MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
- Seq:2530
Specifications
About this item
:
:107.87 g/mol
Merck Index:12,08647
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:962 °C
Density:10.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-131-3
CAS Number:7440-22-4
Formula:Ag
MDL Number:MFCD00003397