Order Entry
Germany
ContactUsLinkComponent
Silver ≥99.9%, wool for elementary analysis, Supelco®
Silver ≥99.9%, wool for elementary analysis, Supelco®
Catalog # 1.01506.0010
Supplier:  Merck
CAS Number:  
Silver ≥99.9%, wool for elementary analysis, Supelco®
Catalog # 1.01506.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Identity:
    passes test
  • Purity:
    ≥ 99.9 %
  • Diameter of fibre:
    about 45 - 55 µm

Specification Test Results

About this item

 :
 :107.87 g/mol
Merck Index:12,08647
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:961.9 °C
Density:10.5 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-131-3
CAS Number:7440-22-4
Formula:Ag
MDL Number:MFCD00003397