Order Entry
Germany
ContactUsLinkComponent
Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.05 mm (0.002 in)
Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.05 mm (0.002 in)
Catalog # 43081.GH
CAS Number:  
Silver ≥99.9% (metals basis), foil, annealed, Thickness: 0.05 mm (0.002 in)
Catalog # 43081.GH
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1736
  • Pk Info:
    100×100 mm

Specifications

About this item

 :
 :107.87 g/mol
Merck Index:12,08647
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:962 °C
Density:10.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-131-3
CAS Number:7440-22-4
Formula:Ag
MDL Number:MFCD00003397