Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silver
Silver ≥99.95% (metals basis), plate, Thickness: 3.175 mm (0.125 in)
Silver ≥99.95% (metals basis), plate, Thickness: 3.175 mm (0.125 in)
:
Thermo Fisher Scientific
Silver ≥99.95% (metals basis), plate, Thickness: 3.175 mm (0.125 in)
:
Thermo Fisher Scientific
:
7440-22-4
:
:
Ag
:
MFCD00003397
:
107.87 g/mol
:
12,08647
:
2212 °C (1013 hPa)
:
962 °C
:
10.5 g/cm³ (20 °C)
:
Ambient
:
231-131-3
msds_ALFA013420
Thermo Scientific Acroseal 25 Years