Order Entry
Germany
ContactUsLinkComponent
Riboflavin 5'-monophosphate sodium salt hydrate
Riboflavin 5'-monophosphate sodium salt hydrate
Supplier:  Cayman Chemical
Certificates
Riboflavin 5'-monophosphate sodium salt hydrate
Supplier:  Cayman Chemical
Certificates

Specifications

  • Storage conditions:
    −20 °C
  • Enzyme name:
    Riboflavin 5'-monophosphate sodium salt hydrate
  • Purity:
    ≥95%
  • Molecular weight:
    514,4 g/mol

Specifications

Related Information