Order Entry
Germany
ContactUsLinkComponent
Aluminium chloride hexahydrate 95.0-101.0% USP, BP, Ph. Eur., pharma grade
Irritant
Aluminium chloride hexahydrate 95.0-101.0% USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates
Irritant
Aluminium chloride hexahydrate 95.0-101.0% USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • Assay (Compl.):
    95.0 - 101.0%
  • Identity According to Pharmacopoeias:
    Passes Test
  • Appearance of solution:
    Passes Test
  • Insoluble matter in H₂O:
    0.0002
  • Ammonium (NH₄):
    0.0005
  • Sulphate (SO₄):
    0.0001
  • Residual solvents (Ph.Eur/:
    Passes Test
  • Alkali and alkaline-earth:
    0.5%
  • Water (H₂O):
    42.0 - 48.0%
  • Heavy metals (as Pb):
    0.002%
  • Iron (Fe):
    0.001%
  • Cadmium (Cd) - ICH Q3D:
    Max. 0.5 ppm
  • Lead (Pb) - ICH Q3D:
    Max. 0.5 ppm
  • Arsenic (As) - ICH Q3D:
    Max. 1.5 ppm
  • Mercury (Hg) - ICH Q3D:
    Max. 1.5 ppm
  • Cobalt (Co) - ICH Q3D:
    Max. 5 ppm
  • Vanadium (V) - ICH Q3D:
    Max. 10 ppm
  • Nickel (Ni) - ICH Q3D:
    Max. 20 ppm
  • Thallium (Tl) - ICH Q3D:
    Max. 0.8 ppm
  • Gold (Au) - ICH Q3D:
    Max. 10 ppm
  • Palladium (Pd) - ICH Q3D:
    Max. 10 ppm
  • Iridium (Ir) - ICH Q3D:
    Max. 10 ppm
  • Osmium (Os) - ICH Q3D:
    Max. 10 ppm
  • Rhodium (Rh) - ICH Q3D:
    Max. 10 ppm
  • Ruthenium (Ru) - ICH Q3D:
    Max. 10 ppm
  • Selenium (Se) - ICH Q3D:
    Max. 15 ppm
  • Silver (Ag) - ICH Q3D:
    Max. 15 ppm
  • Platinum (Pt) - ICH Q3D:
    Max. 10 ppm
  • Lithium (Li) - ICH Q3D:
    Max. 55 ppm
  • Antimony (Sb) - ICH Q3D:
    Max. 120 ppm
  • Barium (Ba) - ICH Q3D:
    Max. 140 ppm
  • Molybdenum (Mo) - ICH Q3D:
    Max. 25 ppm
  • Copper (Cu) - ICH Q3D:
    Max. 250 ppm
  • Tin (Sn) - ICH Q3D:
    Max. 600 ppm
  • Chromium (Cr) - ICH Q3D:
    Max. 25 ppm
  • Solublility in water at 20 °C:
    1.330 g/l
  • pH (50 g/l):
    2.5

Specification Test Results

Related Information

Certificates
Search by Lot Number