Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodine
Iodine, Sigma-Aldrich®
Iodine, Sigma-Aldrich®
:
Merck
Iodine, Sigma-Aldrich®
:
Merck
:
7553-56-2
:
:
I₂
:
MFCD00011355
:
253.81 g/mol
:
13,05036
:
184.3 °C (1013 hPa)
:
114 °C
:
4.9 g/cm³ (20 °C)
:
Ambient
:
3495
:
8,III
:
M-Clarity™ quality level = MQ200
:
231-442-4
:
GHS08
:
GHS07
:
GHS09