Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Selenium, Sigma-Aldrich®

Toxic
Health
Supplier:   Merck
Selenium, Sigma-Aldrich®
Supplier:  Merck
Certificates
Selenium, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:3283
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 128,00 €
    128,00 €
    1 * 50 g
       128,00 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 407,00 €
    407,00 €
    1 * 250 g
       407,00 €