Sulphur
Catalog # EHERC17025000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:32.07 g/mol
Merck Index:13,09059
Boiling Pt:445 °C (1013 hPa)
Melting Pt:113…119 °C
Density:2.36 g/cm³ (20 °C)
Flash Pt:168 °C
Storage Temperature:Ambient
EINECS:231-722-6
UN:1350
ADR:4.1,III
:GHS07
CAS Number:7704-34-9
Formula:S
MDL Number:MFCD00085316