Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphur
Sulphur ≥99.5%, powder -100 mesh, sublimated
Sulphur ≥99.5%, powder -100 mesh, sublimated
:
Thermo Fisher Scientific
Sulphur ≥99.5%, powder -100 mesh, sublimated
:
Thermo Fisher Scientific
:
7704-34-9
:
:
S
:
MFCD00085316
:
32.07 g/mol
:
13,09059
:
445 °C (1013 hPa)
:
113…119 °C
:
2.36 g/cm³ (20 °C)
:
168 °C
:
Ambient
:
231-722-6
:
1350
:
4.1,III
:
GHS02
:
GHS07
ALFA33394.36_TDS_EN
msds_ALFA033394
Thermo Scientific Acroseal 25 Years